MilliporeSigma
  • Arp2 depletion inhibits sheet-like protrusions but not linear protrusions of fibroblasts and lymphocytes.
Arp2 depletion inhibits sheet-like protrusions but not linear protrusions of fibroblasts and lymphocytes.
Cell motility and the cytoskeleton (2008-08-23)
Susan M Nicholson-Dykstra, Henry N Higgs
ABSTRACT
The Arp2/3 complex-mediated assembly and protrusion of a branched actin network at the leading edge occurs during cell migration, although some studies suggest it is not essential. In order to test the role of Arp2/3 complex in leading edge protrusion, Swiss 3T3 fibroblasts and Jurkat T cells were depleted of Arp2 and evaluated for defects in cell morphology and spreading efficiency. Arp2-depleted fibroblasts exhibit severe defects in formation of sheet-like protrusions at early time points of cell spreading, with sheet-like protrusions limited to regions along the length of linear protrusions. However, Arp2-depleted cells are able to spread fully after extended times. Similarly, Arp2-depleted Jurkat T lymphocytes exhibit defects in spreading on anti-CD3. Interphase Jurkats in suspension are covered with large ruffle structures, whereas mitotic Jurkats are covered by finger-like linear protrusions. Arp2-depleted Jurkats exhibit defects in ruffle assembly but not in assembly of mitotic linear protrusions. Similarly, Arp2-depletion has no effect on the highly dynamic linear protrusion of another suspended lymphocyte line. We conclude that Arp2/3 complex plays a significant role in assembly of sheet-like protrusions, especially during early stages of cell spreading, but is not required for assembly of a variety of linear actin-based protrusions.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, reagent grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%