MilliporeSigma
  • Identification of hnRNP-A1 as a pharmacodynamic biomarker of type I PRMT inhibition in blood and tumor tissues.
Identification of hnRNP-A1 as a pharmacodynamic biomarker of type I PRMT inhibition in blood and tumor tissues.
Scientific reports (2020-12-19)
Paul B Noto, Timothy W Sikorski, Francesca Zappacosta, Craig D Wagner, Rocio Montes de Oca, Matthew E Szapacs, Roland S Annan, Yan Liu, Charles F McHugh, Helai P Mohammad, Steven P Piccoli, Caretha L Creasy
ABSTRACT
Arginine methylation has been recognized as a post-translational modification with pleiotropic effects that span from regulation of transcription to metabolic processes that contribute to aberrant cell proliferation and tumorigenesis. This has brought significant attention to the development of therapeutic strategies aimed at blocking the activity of protein arginine methyltransferases (PRMTs), which catalyze the formation of various methylated arginine products on a wide variety of cellular substrates. GSK3368715 is a small molecule inhibitor of type I PRMTs currently in clinical development. Here, we evaluate the effect of type I PRMT inhibition on arginine methylation in normal human peripheral blood mononuclear cells and utilize a broad proteomic approach to identify type I PRMT substrates. This work identified heterogenous nuclear ribonucleoprotein A1 (hnRNP-A1) as a pharmacodynamic biomarker of type I PRMT inhibition. Utilizing targeted mass spectrometry (MS), methods were developed to detect and quantitate changes in methylation of specific arginine residues on hnRNP-A1. This resulted in the development and validation of novel MS and immune assays useful for the assessment of GSK3368715 induced pharmacodynamic effects in blood and tumors that can be applied to GSK3368715 clinical trials.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Ficoll® Paque Plus, Cytiva 17-1440-02, pack of 6 × 100 mL
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%