MilliporeSigma
  • Deficiency of intellectual disability-related gene Brpf1 reduced inhibitory neurotransmission in MGE-derived GABAergic interneurons.
Deficiency of intellectual disability-related gene Brpf1 reduced inhibitory neurotransmission in MGE-derived GABAergic interneurons.
G3 (Bethesda, Md.) (2021-03-22)
Jingli Cao, Weiwei Xian, Maierdan Palihati, Yu Zhu, Guoxiang Wang, Yunli Xie, Guomin Zhou, Linya You
ABSTRACT
Intellectual disability is closely related to impaired GABA neurotransmission. Brpf1 was specifically expressed in medial ganglionic eminence (MGE), a developmental niche of GABAergic interneurons, and patients with BRPF1 mutations showed intellectual disability. To test its role in development and function of MGE-derived GABAergic interneurons, we performed immunofluorescence staining, whole-cell patch-clamp, MGE transplantation and mRNA-Seq to understand its effect on neuronal differentiation, dendritic morphology, electrophysiology, migration and gene regulation, using mouse MGE-derived GABAergic interneurons infected with AAV-shBrpf1. The results showed that Brpf1 knockdown had a decreasing trend, although not significant, on the differentiation of GABAergic interneurons into parvalbumin+ interneurons. Moreover, increased firing threshold, decreased number of evoked action potentials, and a reduced amplitude of miniature inhibitory postsynaptic currents were observed before any significant change of MAP2+ dendritic morphology and in vivo migration ability appeared. Finally, mRNA-Seq analysis revealed that genes related to neurodevelopment and synaptic transmission such as Map2k7 were dysregulated. Our results demonstrated a key role of Brpf1 in inhibitory neurotransmission and related gene expression of GABAergic interneurons.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Monoclonal Anti-GABA antibody produced in mouse, clone GB-69, ascites fluid