MilliporeSigma
  • High Amount of Transcription Factor IRF8 Engages AP1-IRF Composite Elements in Enhancers to Direct Type 1 Conventional Dendritic Cell Identity.
High Amount of Transcription Factor IRF8 Engages AP1-IRF Composite Elements in Enhancers to Direct Type 1 Conventional Dendritic Cell Identity.
Immunity (2020-08-17)
Sunkyung Kim, Prachi Bagadia, David A Anderson, Tian-Tian Liu, Xiao Huang, Derek J Theisen, Kevin W O'Connor, Ray A Ohara, Arifumi Iwata, Theresa L Murphy, Kenneth M Murphy
ABSTRACT
Development and function of conventional dendritic cell (cDC) subsets, cDC1 and cDC2, depend on transcription factors (TFs) IRF8 and IRF4, respectively. Since IRF8 and IRF4 can each interact with TF BATF3 at AP1-IRF composite elements (AICEs) and with TF PU.1 at Ets-IRF composite elements (EICEs), it is unclear how these factors exert divergent actions. Here, we determined the basis for distinct effects of IRF8 and IRF4 in cDC development. Genes expressed commonly by cDC1 and cDC2 used EICE-dependent enhancers that were redundantly activated by low amounts of either IRF4 or IRF8. By contrast, cDC1-specific genes relied on AICE-dependent enhancers, which required high IRF concentrations, but were activated by either IRF4 or IRF8. IRF8 was specifically required only by a minority of cDC1-specific genes, such as Xcr1, which could distinguish between IRF8 and IRF4 DNA-binding domains. Thus, these results explain how BATF3-dependent Irf8 autoactivation underlies emergence of the cDC1-specific transcriptional program.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥90% (HPLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Roche
Collagenase D, from Clostridium histolyticum
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested
Sigma-Aldrich
Poly(deoxyinosinic-deoxycytidylic) acid sodium salt, double-stranded alternating copolymer
Sigma-Aldrich
Aprotinin Nicotiana tobacco, >= 5TIU/mg protein, >= 98 % SDS-PAGE | 9087-70-1, recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, ≥98% (SDS-PAGE)