MilliporeSigma
  • Pro-cachectic factors link experimental and human chronic kidney disease to skeletal muscle wasting programs.
Pro-cachectic factors link experimental and human chronic kidney disease to skeletal muscle wasting programs.
The Journal of clinical investigation (2021-06-02)
Francesca Solagna, Caterina Tezze, Maja T Lindenmeyer, Shun Lu, Guochao Wu, Shuya Liu, Yu Zhao, Robert Mitchell, Charlotte Meyer, Saleh Omairi, Temel Kilic, Andrea Paolini, Olli Ritvos, Arja Pasternack, Antonios Matsakas, Dominik Kylies, Julian Schulze Zur Wiesch, Jan-Eric Turner, Nicola Wanner, Viji Nair, Felix Eichinger, Rajasree Menon, Ina V Martin, Barbara M Klinkhammer, Elion Hoxha, Clemens D Cohen, Pierre-Louis Tharaux, Peter Boor, Tammo Ostendorf, Matthias Kretzler, Marco Sandri, Oliver Kretz, Victor G Puelles, Ketan Patel, Tobias B Huber
ABSTRACT
Skeletal muscle wasting is commonly associated with chronic kidney disease (CKD), resulting in increased morbidity and mortality. However, the link between kidney and muscle function remains poorly understood. Here, we took a complementary interorgan approach to investigate skeletal muscle wasting in CKD. We identified increased production and elevated blood levels of soluble pro-cachectic factors, including activin A, directly linking experimental and human CKD to skeletal muscle wasting programs. Single-cell sequencing data identified the expression of activin A in specific kidney cell populations of fibroblasts and cells of the juxtaglomerular apparatus. We propose that persistent and increased kidney production of pro-cachectic factors, combined with a lack of kidney clearance, facilitates a vicious kidney/muscle signaling cycle, leading to exacerbated blood accumulation and, thereby, skeletal muscle wasting. Systemic pharmacological blockade of activin A using soluble activin receptor type IIB ligand trap as well as muscle-specific adeno-associated virus-mediated downregulation of its receptor ACVR2A/B prevented muscle wasting in different mouse models of experimental CKD, suggesting that activin A is a key factor in CKD-induced cachexia. In summary, we uncovered a crosstalk between kidney and muscle and propose modulation of activin signaling as a potential therapeutic strategy for skeletal muscle wasting in CKD.
Product Description
Roche
DNase I recombinant, grade I, from bovine pancreas, expressed in Pichia pastoris
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00