MilliporeSigma
  • Social isolation-related depression accelerates ethanol intake via microglia-derived neuroinflammation.
Social isolation-related depression accelerates ethanol intake via microglia-derived neuroinflammation.
Science advances (2021-11-06)
Jin-Seok Lee, Sung-Bae Lee, Dong-Woon Kim, Nara Shin, Seon-Ju Jeong, Chae-Ha Yang, Chang-Gue Son
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Supelco
Bicinchoninic Acid solution
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
DAPI, for nucleic acid staining