MilliporeSigma
  • Drug Carriers Based on Graphene Oxide and Hydrogel: Opportunities and Challenges in Infection Control Tested by Amoxicillin Release.
Drug Carriers Based on Graphene Oxide and Hydrogel: Opportunities and Challenges in Infection Control Tested by Amoxicillin Release.
Materials (Basel, Switzerland) (2021-07-03)
Anna Trusek, Edward Kijak
ABSTRACT
Graphene oxide (GO) was proposed as an efficient carrier of antibiotics. The model drug, amoxicillin (AMOX), was attached to GO using a peptide linker (Leu-Leu-Gly). GO-AMOX was dispersed in a hydrogel to which the enzyme responsible for releasing AMOX from GO was also added. The drug molecules were released by enzymatic hydrolysis of the peptide bond in the linker. As the selected enzyme, bromelain, a plant enzyme, was used. The antibacterial nature of the carrier was determined by its ability to inhibit the growth of the Enterococcus faecalis strain, which is one of the bacterial species responsible for periodontal and root canal diseases. The prepared carrier contained only biocompatible substances, and the confirmation of its lack of cytotoxicity was verified based on the mouse fibrosarcoma cell line WEHI 164. The proposed type of preparation, as a universal carrier of many different antibiotic molecules, can be considered as a suitable solution in the treatment of inflammation in dentistry.
Sigma-Aldrich
Artificial Saliva for Pharmaceutical Research
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein