MilliporeSigma
  • Host versus cell-dependent effects of β-arrestin 1 expression in prostate tumorigenesis.
Host versus cell-dependent effects of β-arrestin 1 expression in prostate tumorigenesis.
Carcinogenesis (2021-03-13)
Timothy O Adekoya, Nikia Smith, Ariel J Thomas, Tonya S Lane, Nija Burnette, Elizabeth J Rivers, Yahui Li, Xiaoxin L Chen, Ricardo M Richardson
ABSTRACT
Prostate cancer (PCa) constitutes a serious health challenge and remains one of the main causes of cancer-related death among men. The more aggressive form of the disease has been attributed to androgen independence, resulting in a lack of response to androgen deprivation therapy and sustained activation of other growth pathways. The scaffold proteins β-arrestin 1 and 2 (βarr1 and βarr2), which are known to mediate G protein-coupled receptor desensitization and internalization, were also shown to modulate prostate tumorigenesis. βarr1 is significantly overexpressed (>4-fold) in PCa cells relative to βarr2. In this study, we investigated the effect of βarr1 overexpression in PCa development and progression using the mouse and human PCa cell xenografts, and autochthonous transgenic adenocarcinoma of the mouse prostate (TRAMP) models deficient in β-arrestin depletion of βarr1 in TRAMP mice (TRAMP/βarr1-/-) increased PCa growth and decreased overall survival relative to control TRAMP or TRAMP/βarr2-/- animals. Prostate tissues from TRAMP/βarr1-/- tumors displayed an increase in androgen receptor (AR) expression, whereas overexpression of βarr1 in TRAMP-C1 (TRAMP-C1-βarr1-GFP) which derived from TRAMP decreased AR expression, cell proliferation and tumor growth in nude mice xenografts, relative to control TRAMP-C1-GFP. Knockdown of βarr1 expression in human MDA PCa 2b cells (MDA PCa 2b-βarr1-/-) also decreased AR expression cell proliferation and tumor growth relative to control (MDA PCa 2b-Sham) cells. Interestingly, both TRAMP-C1-βarr1-GFP and MDA PCa 2b-βarr1-/- xenografts showed a decrease in AKT phosphorylation but an increase in MAPK activation. Altogether, the data indicate that the effect of βarr1 in modulating AR signaling to regulate PCa aggressiveness is cell and host autonomous.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture