MilliporeSigma
Blot-MS of Carbonylated Proteins: A Tool to Identify Oxidized Proteins.
Methods in molecular biology (Clifton, N.J.) (2016-09-11)
Rita Ferreira, Pedro Domingues, Francisco Amado, Rui Vitorino
ABSTRACT
The efficiency of proteostasis regulation declines during aging and the failure of protein homeostasis is common in age-related diseases. Protein oxidation is a major contributor to the loss of proteome homeostasis, also called "proteostasis," precluding protein misfolding and aggregation. So, the identification of the molecular pathways impaired by protein oxidation will increase the understanding of proteostasis and the pathophysiological conditions related to the loss of proteostasis. Sample derivatization with dinitrophenyl hydrazine and western blot immunoassay detection of carbonylated proteins (commonly known as Oxyblot™) coupled to mass spectrometry (blot-MS) is an attractive methodological approach to identify proteins that are more prone to carbonylation, a typical oxidative modification of amino acid residues. The integration of blot-MS data of carbonylated proteins with bioinformatics tools allows the identification of the biological processes more affected by protein oxidation and that, eventually, result in the loss of proteostasis.In this chapter, we describe a blot-MS methodology to identify the proteins more prone to oxidation in biological samples, as cell and tissue extracts, and biofluids. Analysis of mitochondria isolated from cardiac tissue is provided as an example. Bioinformatic strategy to deal with data retrieved from blot-MS experiments are proposed for the identification of relevant biological processes modulated by oxidative stress stimuli.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$81.70
$71.08
50 mg
In Stock
Details...
$329.00