MilliporeSigma
  • Enteric nervous system damage caused by abnormal intestinal butyrate metabolism may lead to functional constipation.
Enteric nervous system damage caused by abnormal intestinal butyrate metabolism may lead to functional constipation.
Frontiers in microbiology (2023-05-25)
Le Wang, Wan-Qiang Lv, Jun-Ting Yang, Xu Lin, Hui-Min Liu, Hang-Jing Tan, Ru-Ping Quan, Pan-Pan Long, Hui Shen, Jie Shen, Hong-Wen Deng, Hong-Mei Xiao
ABSTRACT
Functional constipation (FC) is a high morbidity gastrointestinal disease for which dysfunction in the enteric nervous system is a major pathogenesis mechanism. To enhance our understanding of the involvement of intestinal microbiota and its metabolites in the pathogenesis of FC, we conducted a shotgun metagenomic sequencing analysis of gut microbiota and serum short-chain fatty acids (SCFAs) analysis in 460 Chinese women with different defecation frequencies. We observed that the abundance ofFusobacterium_varium, a butyric acid-producing bacterium, was positively correlated (P = 0.0096) with the frequency of defecation; however, the concentrations of serum butyric acid was negatively correlated (P = 3.51E-05) with defecation frequency. These results were verified in an independent cohort (6 patients with FC and 6 controls). To further study the effects of butyric acid on intestinal nerve cells, we treated mouse intestinal neurons in vitro with various concentrations of butyrate (0.1, 0.5, 1, and 2.5 mM). We found that intestinal neurons treated with 0.5 mM butyrate proliferated better than those in the other treatment groups, with significant differences in cell cycle and oxidative phosphorylation signal pathways. We suggest that the decreased butyrate production resulting from the reduced abundance of Fusobacterium in gut microbiota affects the proliferation of intestinal neurons and the energy supply of intestinal cells. However, with FC disease advancing, the consumption and excretion of butyric acid reduce, leading to its accumulation in the intestine. Moreover, the accumulation of an excessively high amount of butyric acid inhibits the proliferation of nerve cells and subsequently exacerbates the disease.
Sigma-Aldrich
Collagenase Type IV, Cls IV
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)