MilliporeSigma
  • Postnatal Zika virus infection leads to morphological and cellular alterations within the neurogenic niche.
Postnatal Zika virus infection leads to morphological and cellular alterations within the neurogenic niche.
Disease models & mechanisms (2024-02-28)
Jéssica C C G Ferreira, Raissa R Christoff, Tailene Rabello, Raiane O Ferreira, Carolina Batista, Pedro Junior Pinheiro Mourão, Átila D Rossi, Luiza M Higa, Maria Bellio, Amilcar Tanuri, Patricia P Garcez
ABSTRACT
The Zika virus received significant attention in 2016, following a declaration by the World Health Organization of an epidemic in the Americas, in which infections were associated with microcephaly. Indeed, prenatal Zika virus infection is detrimental to fetal neural stem cells and can cause premature cell loss and neurodevelopmental abnormalities in newborn infants, collectively described as congenital Zika syndrome. Contrastingly, much less is known about how neonatal infection affects the development of the newborn nervous system. Here, we investigated the development of the dentate gyrus of wild-type mice following intracranial injection of the virus at birth (postnatal day 0). Through this approach, we found that Zika virus infection affected the development of neurogenic regions within the dentate gyrus and caused reactive gliosis, cell death and a decrease in cell proliferation. Such infection also altered volumetric features of the postnatal dentate gyrus. Thus, we found that Zika virus exposure to newborn mice is detrimental to the subgranular zone of the dentate gyrus. These observations offer insight into the cellular mechanisms that underlie the neurological features of congenital Zika syndrome in children.
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse