MilliporeSigma
  • Synthesis of N-substituted acridinediones and polyhydroquinoline derivatives in refluxing water.
Synthesis of N-substituted acridinediones and polyhydroquinoline derivatives in refluxing water.
Molecules (Basel, Switzerland) (2012-05-09)
Jing-Jing Xia, Ke-Hua Zhang
ABSTRACT
Acridinediones were synthesized by the one-pot Hantzsch condensation of an aromatic aldehyde, 5,5-dimethyl-1,3-cyclohexanedione, and aniline/4-methylaniline in refluxing water. This method has then been extended to the four-component reaction of an aromatic aldehyde, 5,5-dimethyl-1,3-cyclohexanedione, ethyl acetoacetate and ammonium acetate for the synthesis of polyhydroquinoline derivatives. This is an environmentally friendly and efficient procedure providing  good to excellent yields.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, 98%
Pricing and availability is not currently available.
Supelco
N-Methylaniline, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetoacetate, puriss. p.a., ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetoacetate, Arxada quality, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetoacetate, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetoacetate, natural, ≥97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetoacetate, ReagentPlus®, 99%
Pricing and availability is not currently available.