Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Pricing and availability is not currently available.
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
Reagent Alcohol, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Pricing and availability is not currently available.
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Pricing and availability is not currently available.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Pricing and availability is not currently available.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and availability is not currently available.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Pricing and availability is not currently available.
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Pricing and availability is not currently available.