MilliporeSigma
  • Isolation of cellulose from rice straw and its conversion into cellulose acetate catalyzed by phosphotungstic acid.
Isolation of cellulose from rice straw and its conversion into cellulose acetate catalyzed by phosphotungstic acid.
Carbohydrate polymers (2013-04-03)
Guozhi Fan, Min Wang, Chongjing Liao, Tao Fang, Jianfen Li, Ronghui Zhou
ABSTRACT
Cellulose was isolated from rice straw by pretreatment with dilute alkaline and acid solutions successively, and it was further transferred into cellulose acetate in the presence of acetic anhydride and phosphotungstic acid (H3PW12O40·6H2O). The removal of hemicellulose and lignin was affected by the concentration of KOH and the immersion time in acetic acid solution, and 83wt.% content of cellulose in the treated rice straw was obtained after pretreatment with 4% KOH and immersion in acetic acid for 5h. Phosphotungstic acid was found to be an effective catalyst for the acetylation of the cellulose derived from rice straw. The degree of substitution (DS) values revealed a significant effect for the solubility of cellulose acetate, and the acetone-soluble cellulose acetate with DS values around 2.2 can be obtained by changing the amount of phosphotungstic acid and the time of acetylation. Both the structure of cellulose separated from rice straw and cellulose acetate were confirmed by FTIR and XRD.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, JIS special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M in ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.02 M in ethanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in isopropanol
SKU
Pack Size
Availability
Price
Quantity
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder