MilliporeSigma
  • Disturbance of redox homeostasis by ornithine and homocitrulline in rat cerebellum: a possible mechanism of cerebellar dysfunction in HHH syndrome.
Disturbance of redox homeostasis by ornithine and homocitrulline in rat cerebellum: a possible mechanism of cerebellar dysfunction in HHH syndrome.
Life sciences (2013-06-29)
Angela Zanatta, Carolina Maso Viegas, Anelise Miotti Tonin, Estela Natacha Brandt Busanello, Mateus Grings, Alana Pimentel Moura, Guilhian Leipnitz, Moacir Wajner
ABSTRACT
Cerebellar ataxia is commonly observed in hyperornithinemia-hyperammonemia-homocitrullinuria (HHH) syndrome, an inherited metabolic disorder biochemically characterized by ornithine (Orn), homocitrulline (Hcit) and ammonia accumulation. Since the pathophysiology of cerebellum damage in this disorder is still unknown, we investigated the effects of Hcit and Orn on important parameters of redox and energy homeostasis in cerebellum of young rats. We determined thiobarbituric acid-reactive substance (TBA-RS) levels, carbonyl content, nitrate and nitrite production, hydrogen peroxide production, GSH concentrations, sulfhydryl content, as well as activities of respiratory chain complexes I-IV, creatine kinase, Na(+),K(+)-ATPase, aconitase and α-ketoglutarate dehydrogenase. Orn and Hcit significantly increased TBA-RS levels (lipid oxidation), that was totally prevented by melatonin and reduced glutathione (GSH). We also found that nitrate and nitrite production was not altered by any of the metabolites, in contrast to hydrogen peroxide production which was significantly enhanced by Hcit. Furthermore, GSH concentrations were significantly reduced by Orn and Hcit and sulfhydryl content by Orn, implying an impairment of antioxidant defenses. As regards energy metabolism, Orn and Hcit provoked a significant reduction of aconitase activity, without altering the other parameters. Furthermore, Orn-elicited reduction of aconitase activity was totally prevented by GSH, indicating that the critical groups of this enzyme were susceptible to oxidation caused by this amino acid. Taken together, our data indicate that redox homeostasis is disturbed by the major metabolites accumulating in HHH syndrome and that this mechanism may be implicated in the ataxia and cerebellar abnormalities observed in this disorder.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Aconitase from porcine heart
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
L-Ornithine monohydrochloride, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioXtra, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
L-Ornithine hydrochloride, 99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
L-Ornithine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
L-Ornithine monohydrochloride, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
DL-Ornithine monohydrochloride, ≥99.0% (AT)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Citrulline, ≥98% (TLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%