MilliporeSigma
[Chemical constituents of Salvia chinensis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-08-21)
Jun-Feng Gao, Ling Ding, Peng Zhang, Jian-Xun Liu
ABSTRACT
Chemical constituents were investigated on the ethyl acetate extract of Salvia chinensis. Compounds were separated and purified by various chromatograhic techniques including silica gel, Sephadex LH-20 and reversed-phase HPLC. Their structures were identified by spectroscopic data analysis. Eleven compounds were isolated and purified and their structures were identified as oresbiusin A(1), ethyl dihydrocaffeate (2), ethyl rosmarinate (3), rosmarinic acid (4), methyl rosmarinate (5), bis (2-ethylhexyl) phthalate (6), salvianol acid C (7), methyl salvianol acid C (8), methyl salvianolate A (9), dimethyl lithospermate B (10), and salvianolic acid A(11). Except for rosmarinic acid, the remaining compounds were isolated from S. chinensis for the first time.
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, fine
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Sephadex® G-50, Superfine
Supelco
Sephadex® G-50, Medium
Sigma-Aldrich
Sephadex® G-50, Fine
Supelco
Salvianolic acid A, analytical standard
Supelco
Dextran, analytical standard, for GPC, 25,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Sigma-Aldrich
Rosmarinic acid, 96%
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-25, Medium
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Sephadex® G-25, Fine