MilliporeSigma
  • Simultaneous preconcentration and determination of Ni and Pb in water samples by solid-phase extraction and flame Atomic absorption spectrometry.
Simultaneous preconcentration and determination of Ni and Pb in water samples by solid-phase extraction and flame Atomic absorption spectrometry.
Journal of AOAC International (2013-09-05)
Harun Ciftci, Mehmet Mursit Temuz, Esen Ciftci
ABSTRACT
An analytical method for the preconcentration of nickel (Ni) and lead (Pb) in water samples on polystyrene-graft-ethylmethacrylate copolymer resin has been developed. The determinations of metal ions were performed by flame atomic absorption spectrometry. The experimental parameters, such as solution pH, sample flow rate, type and concentration of eluent, and amount of resin, were optimized for the preconcentration and recovery of Ni and Pb ions. Adsorbed ions on the resin were eluted by 6 mL of 2 M HNO3. Under optimized conditions, the LOD was found to be 0.92 and 1.44 microg/L for Ni and Pb ions, respectively. The accuracy of the proposed procedure was tested by recovery studies and analysis of a standard reference material (GBW 08301, river sediment). The method was successfully applied to water samples.
Product Description
Supelco
Water, for TOC analysis
Sigma-Aldrich
Distilled water, suitable for HPLC
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis