Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing Supelco
Palmitic acid, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hippuric acid, 98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
cis-Aconitic acid, ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Palmitic acid, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sign Into View Organizational & Contract Pricing Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sign Into View Organizational & Contract Pricing Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sign Into View Organizational & Contract Pricing Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Supelco
Glutaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaric acid, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sign Into View Organizational & Contract Pricing