Sigma-Aldrich
Triethylamine, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing Supelco
Acetone, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, natural, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing