MilliporeSigma
  • Zinc-L-carnosine binds to molecular chaperone HSP70 and inhibits the chaperone activity of the protein.
Zinc-L-carnosine binds to molecular chaperone HSP70 and inhibits the chaperone activity of the protein.
Journal of biochemistry (2013-05-21)
Asami Haga, Tomoya Okamoto, Shintaroh Yamada, Toshihiko Kubota, Ann Sanpei, Shota Takahashi, Masahiro Nakayama, Miki Nagai, Michiro Otaka, Toshio Miyazaki, Wataru Nunomura, Ewa Grave, Hideaki Itoh
ABSTRACT
In this study, we have investigated the specific binding proteins of Zinc-L-carnosine (Polaprezinc) using Polaprezinc-affinity column chromatography in vitro. A protein having a 70-kDa molecular mass was eluted by the linear gradient of 0-1.0 mM Polaprezinc from the affinity column and the protein was identified as the molecular chaperone HSP70 by immunoblotting. The chaperone activity of HSP70 was completely suppressed by Polaprezinc. The ATPase activity of HSP70 was affected to some extent by the reagent. In the circular dichroism (CD) spectrum, the secondary structure of HSP70 was changed in the presence of Polaprezinc, i.e. it decreased in the α-helix. We have determined the Polaprezinc-binding domain of HSP70 by using recombinant HSP70N- and C-domains. Although Polaprezinc could bind to both the N-terminal and the C-terminal of HSP70, the HSP70N-domain has a high affinity to the drug. Regarding the peptide cleavage of the HSP70N- and C-domains with proteinase K, the intact HSP70N still remained in the presence of Polaprezinc. On the other hand, the quantity of the intact C-domain slightly decreased under the same conditions along with the newly digested small peptides appeared. It has been suggested that Polaprezinc binds to HSP70 especially in the N-domains, suppresses the chaperone activity and delays an ATPase activities of HSP70.
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
L-Carnosine, ~99%, crystalline
Sigma-Aldrich
Proteinase K from Tritirachium album, Reagents designed and manufactured under current ISO 13485 certification.