MilliporeSigma
  • A highly electron-deficient analogue of aniline, soluble oligomers, and their redox properties.
A highly electron-deficient analogue of aniline, soluble oligomers, and their redox properties.
The Journal of organic chemistry (2013-08-27)
Brandon Djukic, Alan J Lough, Dwight S Seferos
ABSTRACT
The synthesis and electrochemical oxidative coupling of a highly electron-deficient analogue of aniline results in the formation of soluble electron-deficient oligomers. Oligomers undergo related oxidation and reduction processes that are separated by a wide potential range. The mechanism behind this behavior is examined by cyclic voltammetry, optical absorption spectroscopy, (1)H NMR spectroscopy, and density functional theory calculations. Mesomeric isomerization of the oxidized oligomers leads to a very stable oxidized state that requires a large (2.8 V) overpotential to return to the neutral form.
Product Description
Supelco
Aniline, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 24, 2025
$154.00
500 mL
Estimated to ship on March 24, 2025
$522.00