MilliporeSigma
  • Refrigeration provides a simple means to synchronize in vitro cultures of Plasmodium falciparum.
Refrigeration provides a simple means to synchronize in vitro cultures of Plasmodium falciparum.
Experimental parasitology (2014-03-19)
Lili Yuan, Mingming Hao, Lanou Wu, Zhen Zhao, Benjamin M Rosenthal, Xiaomei Li, Yongshu He, Ling Sun, Guohua Feng, Zheng Xiang, Liwang Cui, Zhaoqing Yang
ABSTRACT
Plasmodium falciparum is usually asynchronous during in vitro culture. Highly synchronized cultures of P. falciparum are routinely used in malaria research. Here, we describe a simple synchronization procedure for P. falciparum asexual erythrocytic culture, which involves storage at 4°C for 8-24 h followed by routine culture. When cultures with 27-60% of ring stage were synchronized using this procedure, 70-93% ring stages were obtained after 48 h of culture and relative growth synchrony remained for at least two erythrocytic cycles. To test the suitability of this procedure for subsequent work, drug sensitivity assays were performed using four laboratory strains and four freshly adapted clinical P. falciparum isolates. Parasites synchronized by sorbitol treatment or refrigeration showed similar dose-response curves and comparable IC50 values to four antimalarial drugs. The refrigeration synchronization method is simple, inexpensive, time-saving, and should be especially useful when large numbers of P. falciparum culture are handled.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, FCC, FG
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)