MilliporeSigma
  • Dimorphism and hydrocarbon metabolism in Yarrowia lipolytica var. indica.
Dimorphism and hydrocarbon metabolism in Yarrowia lipolytica var. indica.
Archives of microbiology (2014-05-21)
A S Palande, S V Kulkarni, C León-Ramirez, E Campos-Góngora, J Ruiz-Herrera, M V Deshpande
ABSTRACT
Yarrowia lipolytica is able to metabolize high Mr hydrophobic natural compounds such as fatty acids and hydrocarbons. Characteristically, strains of Y. lipolytica can grow as populations with variable proportions of yeast and filamentous forms. In the present study, we describe the dimorphic characteristics of a variant designated as Y. lipolytica var. indica isolated from petroleum contaminated sea water and the effect of cell morphology on hydrocarbon metabolism. The variant behaved as a yeast monomorphic strain, under conditions at which terrestrial Y. lipolytica strain W29 and its derived strains, grow as almost uniform populations of mycelial cells. Using organic nitrogen sources and N-acetylglucosamine as carbon source, var. indica was able to form mycelial cells, the proportion of which increased when incubated under semi-anaerobic conditions. The cell surface characteristics of var. indica and W29 were found to be different with respect to contact angle and percent hydrophobicity. For instance, percent hydrophobicity of var. indica was 89.93 ± 1.95 while that of W29 was 70.78 ± 1.1. Furthermore, while all tested strains metabolize hydrocarbons, only var. indica was able to use it as a carbon source. Yeast cells of var. indica metabolized hexadecane with higher efficiency than the mycelial form, whereas the mycelial form of the terrestrial strain metabolized the hydrocarbon more efficiently, as occurred with the mycelial monomorphic mutant AC11, compared to the yeast monomorphic mutant AC1.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
Hexadecane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Nonne-Apelt′s reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, ≥99.5%, for biological purposes
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
SAFC
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$31.50
5 mL
In Stock
Details...
$122.00
$113.46