MilliporeSigma
  • Cod liver oil ameliorates sodium nitrite-induced insulin resistance and degradation of rat hepatic glycogen through inhibition of cAMP/PKA pathway.
Cod liver oil ameliorates sodium nitrite-induced insulin resistance and degradation of rat hepatic glycogen through inhibition of cAMP/PKA pathway.
Life sciences (2014-12-03)
Mohammed M H Al-Gayyar, Abdullah Alyoussef, Ahmed M Hamdan, Ahmed Abbas, Mohamed M Darweish, Amany A El-Hawwary
ABSTRACT
Sodium nitrite is used to inhibit the growth of microorganisms and is responsible for the desirable red color of meat; however, it can be toxic in high quantities for humans and other animals. Moreover, glycogen, a branched polysaccharide, efficiently stores and releases glucose monosaccharides to be accessible for metabolic and synthetic requirements of the cell. Therefore, we examined the impact of dietary sodium nitrite and cod liver oil on liver glycogen. Thirty-two Sprague-Dawley rats were treated daily with sodium nitrite (80 mg/kg) in the presence/absence of cod liver oil (5 ml/kg). Liver sections were stained with Periodic acid-Schiff. Hepatic homogenates were used for measurements of glycogen, cyclic adenosine monophosphate (cAMP), protein kinase A (PKA), glycogen synthase, glycogen synthase kinase, pyruvate carboxylase, fructose 1,6-diphosphatase, glucose 6-phosphatase, phosphodiesterase and glycogen phosphorylase. Glucose, pyruvate tolerances and HOMA insulin resistance were also determined. Sodium nitrite significantly increased plasma glucose and insulin resistance. Moreover, sodium nitrite significantly reduced hepatic glycogen content as well as activities of glycogen synthase, glycogen synthase kinase-3, and phosphodiesterase. Sodium nitrite elevated hepatic cAMP, PKA, pyruvate carboxylase, fructose 1,6-diphosphatase, glucose 6-phosphatase and phosphorylase. Cod liver oil significantly blocked all of these except pyruvate carboxylase, fructose 1,6-diphosphatase and glucose 6-phosphatase. Sodium nitrite inhibited liver glycogenesis and enhanced liver glycogenolysis and gluconeogenesis, which is accompanied by hyperglycemia and insulin resistance through the activation of cAMP/PKA and the inhibition of phosphodiesterase. Cod liver oil blocked the sodium nitrite effects on glycogenesis and glycogenolysis without affecting gluconeogenesis.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%