MilliporeSigma
  • Down-regulation of MSH2 expression by Hsp90 inhibition enhances cytotoxicity affected by tamoxifen in human lung cancer cells.
Down-regulation of MSH2 expression by Hsp90 inhibition enhances cytotoxicity affected by tamoxifen in human lung cancer cells.
Biochemical and biophysical research communications (2014-12-10)
Jen-Chung Ko, Hsien-Chun Chiu, Jhan-Jhang Syu, Chien-Yu Chen, Yun-Ting Jian, Yi-Jhen Huang, Ting-Yu Wo, Yi-Jun Jian, Po-Yuan Chang, Tai-Jing Wang, Yun-Wei Lin
ABSTRACT
The anti-estrogen tamoxifen has been used worldwide as an adjuvant hormone therapeutic agent in the treatment of breast cancer. However, the molecular mechanism of tamoxifen-induced cytotoxicity in non-small cell lung cancer (NSCLC) cells has not been identified. Human MutS homolog 2 (MSH2), a crucial element of the highly conserved DNA mismatch repair system, and expression of MSH2 have been down-regulated by Hsp90 function inhibition in human lung cancer. Therefore, in this study, we examined whether MSH2 plays a role in the tamoxifen and Hsp90 inhibitor-induced cytotoxic effect on NSCLC cells. The results showed that treatment with tamoxifen increased MSH2 mRNA and protein levels. The combination treatment with PI3K inhibitors (LY294002 or wortmannin) or knockdown AKT expression by specific small interfering RNA could decrease tamoxifen-induced MSH2 expression. Both knocking down MSH2 expression and co-treatment of PI3K inhibitors enhanced the cytotoxicity and cell growth inhibition of tamoxifen. Compared to a single agent alone, tamoxifen combined with an Hsp90 inhibitor resulted in cytotoxicity and cell growth inhibition synergistically in NSCLC cells, accompanied with reduced MSH2 expression. These findings may have implications for the rational design of future drug regimens incorporating tamoxifen and Hsp90 inhibitors for the treatment of NSCLC.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Tamoxifen, ≥99%
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Supelco
Tamoxifen, analytical standard