MilliporeSigma
Twenty years of acanthamoeba diagnostics in Austria.
The Journal of eukaryotic microbiology (2014-07-23)
Julia Walochnik, Ute Scheikl, Eva-Maria Haller-Schober
ABSTRACT
Acanthamoebae are the causative agents of an often seriously progressing keratitis (AK) occurring predominantly in contact lens wearers and can cause several disseminating infections potentially resulting in granulomatous amoebic encephalitis (GAE) in the immunocompromised host. Our institution is the Austrian reference laboratory for Acanthamoeba diagnostics and the aim of this study was to give an overview of proven cases of Acanthamoeba infections in Austria during the past 20 yr. All samples of patients with suspected AK or GAE were screened for Acanthamoeba spp. by culture and/or PCR and the detected amoebae were genotyped. Altogether, 154 cases of AK and three cases of GAE were diagnosed. Age of the AK patients ranged from 8 to 82 yr (mean 37.8) and 58% of the patients were female. Approximately 89% of the AK patients were contact lens wearers, almost all cases were unilateral and 19% of the patients required a keratoplasty. Age of the GAE patients ranged from 2 to 25 yr (mean 14.7), all were HIV-negative, but two were severely immunosuppressed at the time of diagnosis. The predominant genotype in the AK cases was T4, other genotypes found were T3, T5, T6, T10 and T11. The three GAE cases involved genotypes T2, T4 and T5.
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
3-Methyl-1-butanol, JIS special grade, ≥98.0%
Sigma-Aldrich
3-Methyl-1-butanol, SAJ first grade, ≥96.0%
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
3-Methylbutanol, for molecular biology, BioReagent, ≥98.5%
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)