MilliporeSigma
Absolute bioavailability and metabolism of aceclofenac in rats.
Archives of pharmacal research (2014-03-19)
Keumhan Noh, Beom Soo Shin, Kwang-il Kwon, Hwi-yeol Yun, Eunyoung Kim, Tae Cheon Jeong, Wonku Kang
ABSTRACT
Aceclofenac is one of the most popular analgesic and anti-inflammatory drugs used for the relief of pain, rheumatoid arthritis, and osteoarthritis. To date, no intravenous preparation of aceclofenac has been developed because of its poor water solubility. In this study, to investigate its absolute bioavailability and metabolism in rats, aceclofenac was dissolved in a sterile aqueous solution containing urea (20 %) and trisodium citrate (10 %), and administered via oral (20 mg/kg) and intravenous (10 mg/kg) routes. Blood samples were taken serially, and aceclofenac and its three major metabolites (4'-hydroxydiclofenac, 4'-hydroxyaceclofenac, and diclofenac) were measured by HPLC-MS/MS. The absolute oral bioavailability of aceclofenac was approximately 15 %. Diclofenac and 4'-hydroxydiclofenac were the main metabolites in rats, in contrast to 4'-hydroxyaceclofenac in humans. The low bioavailability of aceclofenac is likely due to extensive metabolism, and bioavailability may be even lower if the drug were administered as a tablet, considering its low water solubility. This study provides complete time profiles of the plasma concentrations of aceclofenac and its metabolites in rats and highlights the difference in drug metabolism between rats and humans.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Aceclofenac, ≥98% (HPLC)
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Supelco
Urea, analytical standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets