MilliporeSigma
  • Highly sensitive analysis of nucleic acids using capillary gel electrophoresis with ultraviolet detection based on the combination of matrix field-amplified and head-column field-amplified stacking injection.
Highly sensitive analysis of nucleic acids using capillary gel electrophoresis with ultraviolet detection based on the combination of matrix field-amplified and head-column field-amplified stacking injection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Dong-Sheng Lian, Shu-Jin Zhao
ABSTRACT
To develop a highly sensitive method for analyzing nucleic acids using capillary gel electrophoresis with ultraviolet detection (CGE-UV), we combined matrix field-amplified with head-column field-amplified stacking injection (C-FASI) to employ the advantages of two methods. Without diminishing the resolution, a limit of detection of 0.13 ng/ml (signal/noise=3) in a 300,000-fold diluted sample was obtained, the sensitivity is 102,308 times higher than that achieved with normal pressure injection, 3077 times that with normal electrokinetic injection, 154 times that with pressure field-amplified sample stacking injection, and 31 times that with matrix field-amplified stacking injection. After establishing the method, we tested the detection of a φX174-Hae III digest DNA product without purification and with a high ionic strength. At the lowest dilution of 5000-fold, sample at a concentration of 10 ng/ml was enriched and detected. The relative standard deviations for migration time and peak area (n=3) were 0.03-1.15 and 0.72-6.42, respectively. To further validate C-FASI was applicable for real sample, a 400 bp PCR product without purification was directly detected with a limit of detection at the concentration of 6000-fold dilution (signal/noise=3), The relative standard deviations for migration time and peak area (n=6) were 0.44 and 4.8, respectively. These results indicated that C-FASI had good qualitative and quantitative detection abilities and CGE-UV based on C-FASI is easy to perform, practical, highly-sensitive and robust for nucleic acid detection, which makes it a highly valuable tool for genetic diagnostics based on nucleic acid analysis.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%