MilliporeSigma
  • Effects of pentylenetetrazole kindling on mitogen-activated protein kinases levels in neocortex and hippocampus of mice.
Effects of pentylenetetrazole kindling on mitogen-activated protein kinases levels in neocortex and hippocampus of mice.
Neurochemical research (2014-10-16)
Juliana Ben, Paulo Alexandre de Oliveira, Filipe Marques Gonçalves, Tanara Vieira Peres, Filipe Carvalho Matheus, Alexandre Ademar Hoeller, Rodrigo Bainy Leal, Roger Walz, Rui Daniel Prediger
ABSTRACT
The epileptogenesis process involves cell signaling events associated with neuroplasticity. The mitogen-activated protein kinases (MAPKs) integrate signals originating from a variety of extracellular stimuli and may regulate cell differentiation, survival, cell death and synaptic plasticity. Here we compared the total and phosphorylated MAPKs (ERK1/2, JNK1/2 and p38(MAPK)) levels in the neocortex and hippocampus of adult Swiss male mice quantified by western blotting analysis 48 h after the last injection of pentylenetetrazole (PTZ), according to the kindling protocol (35 mg/kg, i.p., on alternated days, with a total of eight injections). The total levels of the investigated MAPKs and the phospho-p38(MAPK) in the neocortex and hippocampus were not affected by the PTZ injections. The MAPKs phosphorylation levels remain unaltered in PTZ-treated animals without convulsive seizures. The phospho-JNK2 phosphorylation, but not the phospho-JNK1, was increased in the hippocampus of PTZ-treated animals showing 1-3 days with convulsive seizures, whereas no significant changes were observed in those animals with more than 3 days with convulsive seizures. The phospho-ERK1/2 phosphorylation decreased in the neocortex and increased in the hippocampus of animals with 1-4 days with convulsive seizures and became unaltered in mice that showed convulsive seizures for more than 4 days. These findings indicate that resistance to PTZ kindling is associated with unaltered ERK1/2, JNK1/2 and p38(MAPK) phosphorylation levels in the neocortex and hippocampus. Moreover, when the PTZ kindling-induced epileptogenesis manifests behaviorally, the activation of the different MAPKs sub-families shows a variable and non-linear pattern in the neocortex and hippocampus.
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.0%
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Luminol, ≥97% (HPLC)