MilliporeSigma
  • Silver dopants for targeted and untargeted direct analysis of unsaturated lipids via infrared matrix-assisted laser desorption electrospray ionization (IR-MALDESI).
Silver dopants for targeted and untargeted direct analysis of unsaturated lipids via infrared matrix-assisted laser desorption electrospray ionization (IR-MALDESI).
Rapid communications in mass spectrometry : RCM (2014-10-11)
Florian Meier, Kenneth P Garrard, David C Muddiman
ABSTRACT
Unsaturated lipids play a crucial role in cellular processes as signaling factors, membrane building blocks or energy storage molecules. However, adequate mass spectrometry imaging of this diverse group of molecules remains challenging. In this study we implemented silver cationization for direct analysis by infrared matrix-assisted laser desorption electrospray ionization (IR-MALDESI) to enhance the ion abundances for olefinic lipids and facilitate peak assignment. Trace amounts of silver nitrate were doped into the electrospray solvent of an IR-MALDESI imaging source coupled to an Orbitrap mass analyzer. Calcifediol was examined as a model compound to demonstrate the effect of silver dopants on sensitivity and assay robustness. Dried human serum spots were subsequently analyzed to compare Ag-doped solvents with previously described solvent compositions. Mass differences as well as ion abundance ratio filters were employed to interpret results based on the characteristic isotopic pattern of silver. Olefinic lipids were readily observed as silver adducts in IR-MALDESI analyses. Silver cationization decreased the limit of detection for calcifediol by at least one order of magnitude and was not affected in complex biological matrices. The ion abundance ratio and mass difference of [M + (107) Ag(+)](+) and [M + (109) Ag(+)](+) were successfully applied to facilitate the spectral assignment of silver adducts. Overall, silver cationization increased the analyte coverage in human serum by 43% compared with a standard IR-MALDESI approach. Silver cationization has been shown to enhance IR-MALDESI sensitivity and selectivity for unsaturated lipids, even when applied to complex samples. Increased compound coverage, enhanced robustness as well as the developed tools for peak assignment and mapping of isotopic patterns will clearly benefit future mass spectrometry imaging studies.
Product Description
Supelco
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Calcifediol, European Pharmacopoeia (EP) Reference Standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Supelco
Oleic acid, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%