MilliporeSigma
  • POF1B localizes to desmosomes and regulates cell adhesion in human intestinal and keratinocyte cell lines.
POF1B localizes to desmosomes and regulates cell adhesion in human intestinal and keratinocyte cell lines.
The Journal of investigative dermatology (2014-08-02)
Arianna Crespi, Alessandra Bertoni, Ilaria Ferrari, Valeria Padovano, Pamela Della Mina, Emilio Berti, Antonello Villa, Grazia Pietrini
ABSTRACT
By means of morphological and biochemical criteria, we here provide evidence for the localization and function of premature ovarian failure, 1B (POF1B) in desmosomes. In monolayers of Caco-2 intestinal cells and in stratified HaCaT keratinocytes, endogenous POF1B colocalized with desmoplakin at desmosome plaques and in cytoplasmic particles aligned along intermediate filaments (IFs). POF1B predominantly co-fractionated with desmosomes and IF components and exhibited properties characteristic of desmosomes (i.e., detergent insolubility and calcium independence). The role of NH2 and COOH domains in the association of POF1B with desmosomes and IFs was revealed by transient expression of the truncated protein in Caco-2 cells and in cells lacking desmosomes. The function of POF1B in desmosomes was investigated in HaCaT keratinocytes stably downregulated for POF1B expression. Transmission electron microscopy analysis revealed a decrease in desmosome number and size, and desmosomes of the downregulated keratinocytes displayed weak electron-dense plaques. Desmosome alterations were associated with defects in cell adhesion, as revealed by the reduced resistance to mechanical stress in the dispase fragmentation assay. Moreover, desmosome localization of POF1B was restricted to granular layers in human healthy epidermis, whereas it largely increased in hyperproliferative human skin diseases, thus demonstrating the localization of POF1B also in desmosomes of multistratified epithelia.
Product Description
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Monoclonal Anti-Cytokeratin Peptide 8 antibody produced in mouse, clone M20, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Plakoglobin (Catenin γ) antibody produced in mouse, clone 15F11, ascites fluid, buffered aqueous solution
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Monoclonal Anti-Cytokeratin Peptide 14 antibody produced in mouse, clone CKB1, ascites fluid
Sigma-Aldrich
G 418 disulfate salt, potency: ≥720 μg per mg (dried basis)
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture