MilliporeSigma
  • Development of stability-indicating UHPLC method for the quantitative determination of silodosin and its related substances.
Development of stability-indicating UHPLC method for the quantitative determination of silodosin and its related substances.
Journal of chromatographic science (2013-07-13)
Jafer Vali Shaik, Shantikumar Saladi, Shakil S Sait
ABSTRACT
A novel, specific and stability-indicating reversed-phase (RP) ultra-high-performance liquid chromatography (UHPLC) method, which is mass compatible, was developed and validated for the quantitative determination of silodosin and its related substances. Silodosin was subjected to stress conditions like hydrolysis (acid and basic), oxidation, photolysis and thermal degradation, as per the guidelines of the International Conference Harmonization, to show that the method is stability-indicating. The proposed UHPLC method has a resolution of greater than 2.0 between silodosin and its process-related impurities. The chromatographic separation was achieved on an Agilent Poroshell 120 EC-C18 column (50 × 4.6mm i.d.; particle size, 2.7 µm). The method employed a linear gradient elution using a mobile phase consisting of acetonitrile and 10 mM ammonium acetate buffer with 0.1% triethyl amine, with pH adjusted to 6.0, monitored at 273 nm. The developed RP-LC method was validated with respect to linearity, accuracy, precision and robustness. The known process impurities were separated and their structure was confirmed by using liquid chromatography-mass spectrometry and direct mass analysis.
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%