MilliporeSigma
  • Effects of prolonged neuronal nitric oxide synthase inhibition on the development and expression of L-DOPA-induced dyskinesia in 6-OHDA-lesioned rats.
Effects of prolonged neuronal nitric oxide synthase inhibition on the development and expression of L-DOPA-induced dyskinesia in 6-OHDA-lesioned rats.
Neuropharmacology (2014-09-10)
Fernando Eduardo Padovan-Neto, Roberta Cavalcanti-Kiwiatkoviski, Ruither Oliveira Gomes Carolino, Janete Anselmo-Franci, Elaine Del Bel
ABSTRACT
It is well known that nitric oxide (NO) interacts with dopamine (DA) within the striatal circuitry. The anti-dyskinetic properties of NO synthase (NOS) inhibitors demonstrate the importance of NO in L-3,4-dihydroxyphenylalanine (L-DOPA)-induced dyskinesia (LID). Here, we investigated the ability of a daily co-treatment of the preferential neuronal NOS (nNOS) inhibitor, 7-nitroindazole (7-NI, 30 mg/kg), with L-DOPA (30 mg/kg) to counteract LID in unilaterally 6-OHDA-lesioned rats. We analyzed striatal nNOS-expressing interneurons, DA and 5-HT neurochemistry in the striatum and alterations of the Fos-B/ΔFosB expression in the corticostriatal, nigrostriatal and mesolimbic pathways. Prolonged administration of 7-NI inhibited the manifestation of chronic L-DOPA treatment-induced abnormal involuntary movements (AIMs). LID was associated with an up-regulation in the number of nNOS-expressing interneurons in the lateral but not medial striatum. nNOS inhibition reduced the number of nNOS-expressing interneurons. The anti-dyskinetic effects of 7-NI correlated with a reduction in DA and 5-HT turnover in the striatum. At postsynaptic striatal sites, 7-NI prevented L-DOPA-induced Fos-B/ΔFosB up-regulation in the motor cortex, nucleus accumbens and striatum. Finally, 7-NI blocked Fos-B/ΔFosB expression in nicotinamide adenine dinucleotide phosphate diaphorase (NADPH-d)-positive interneurons in the striatum. These results provide further evidence of the molecular mechanisms by which NOS-inhibiting compounds attenuate LID. The involvement of NO with DA and 5-HT neurochemistry may contribute to the understanding of this new, non-dopaminergic therapy for the management of LID.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
7-Nitroindazole, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
R-(−)-Apomorphine hydrochloride hemihydrate, calcined, ≥98% (with NaOH, titration)