MilliporeSigma
  • Physiopathological effects of Escherichia coli O157:H7 inoculation in weaned calves fed with colostrum containing antibodies to EspB and Intimin.
Physiopathological effects of Escherichia coli O157:H7 inoculation in weaned calves fed with colostrum containing antibodies to EspB and Intimin.
Vaccine (2014-05-07)
B C Rabinovitz, D A Vilte, M Larzábal, A Abdala, R Galarza, E Zotta, C Ibarra, E C Mercado, A Cataldi
ABSTRACT
Escherichia coli O157:H7 is responsible for severe intestinal disease and hemolytic uremic syndrome (HUS), a serious systemic complication which particularly affects children. Cattle are the primary reservoir for E. coli O157:H7 and the main source of infection for humans. In this study, we evaluated the ability of transferred maternal colostral antibodies against γ-Intimin C280 and EspB, to protect young weaned calves from E. coli O157:H7 infection. Hyperimmune colostra were obtained by immunization of pregnant cows with a mix of the mentioned antigens. All vaccinated cows mounted a significant IgG response against γ-Intimin C280, and EspB in sera and colostra. Colostrum-fed calves also exhibited high serum IgG titers against γ-Intimin C280 and EspB along with a rise in mucosal γ-Intimin C280-specific IgG antibodies at recto-anal junction and ileum. Additionally, 70 day-old calves received a challenge with E. coli O157:H7 but no reduction in total bacterial shedding or frequency of E. coli O157:H7 excretion from these calves was observed. Most tissue samples showed granulocyte focal infiltrations of the lamina propria and enterocyte erosion. In conclusion, up to the 70th day, the passively acquired γ-Intimin-C280 and EspB-IgG antibodies present in sera and recto-anal mucosa reached a titer insufficient to reduce EHEC O157 shedding and damages of experimentally inoculated young calves.
Product Description
Sorbitol, European Pharmacopoeia (EP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, FCC, FG
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)