MilliporeSigma
  • Antibacterial activity of enrofloxacin and ciprofloxacin derivatives of β-octaarginine.
Antibacterial activity of enrofloxacin and ciprofloxacin derivatives of β-octaarginine.
Chemistry & biodiversity (2015-02-14)
Nirupam Purkayastha, Stefania Capone, Albert K Beck, Dieter Seebach, Jennifer Leeds, Katherine Thompson, Heinz E Moser
ABSTRACT
β(3) -Octaarginine chains were attached to the functional groups NH and CO2 H of the antibacterial fluoroquinolones ciprofloxacin (→1) and enrofloxacin (→2), respectively, in order to find out whether the activity increases by attachment of the polycationic, cell-penetrating peptide (CPP) moiety. For comparison, simple amides, 3-5, of the two antimicrobial compounds and β(3) -octaarginine amide (βR8 ) were included in the antibacterial susceptibility tests to clarify the impact of chemical modification on the microbiological activity of either scaffold (Table).
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, for TOC analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, suitable for ion chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, BioPerformance Certified
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, ACS reagent, for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
E-Toxate Water, endotoxin, free
SKU
Pack Size
Availability
Price
Quantity
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Deoxycorticosterone acetate
SKU
Pack Size
Availability
Price
Quantity
Supelco
Diethyl ether, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for molecular biology, sterile filtered
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, HPLC Plus