MilliporeSigma
  • Inutero exposure to diisononyl phthalate caused testicular dysgenesis of rat fetal testis.
Inutero exposure to diisononyl phthalate caused testicular dysgenesis of rat fetal testis.
Toxicology letters (2014-12-03)
Linxi Li, Tiao Bu, Huina Su, Zhichuan Chen, Yuyuan Liang, Gaolong Zhang, Danyan Zhu, Yuanyuan Shan, Renai Xu, Yuanyuan Hu, Junwei Li, Guoxin Hu, Qingquan Lian, Ren-Shan Ge
ABSTRACT
Diisononyl phthalate (DINP) is a synthetic material that has been widely used as a substitute for other plasticizers prohibited due to reproductive toxicity in consumer products. Some phthalates have been associated with testicular dysgenesis syndrome in male fetus when female pregnant dams were exposed to them. The present study investigated effects of DINP on fetal Leydig cell function and testis development. Female pregnant Sprague Dawley rats received control vehicle (corn oil) or DINP (10, 100, 500, and 1000 mg/kg) by oral gavage from gestational day (GD) 12 to 21. At GD 21.5, testicular testosterone production, fetal Leydig cell numbers and distribution, testicular gene and protein expression levels were examined. DINP showed dose-dependent increase of fetal Leydig cell aggregation with the low observed adverse-effect level (LOAEL) of 10 mg/kg and multinucleated gonocyte with LOAEL of 100 mg/kg. At 10 mg/kg, DINP also significantly increased fetal Leydig cell size, but inhibited insulin-like 3 and 3β-hydroxysteroid dehydrogenase gene expression and protein levels. DINP inhibited testicular testosterone levels at 1000 mg/kg. The results indicate that in utero exposure to DINP affects the expression levels of some fetal Leydig cell steroidogenic genes, gonocyte multinucleation and Leydig cell aggregation.
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)