MilliporeSigma
  • Genetic analysis of an allergic rhinitis cohort reveals an intercellular epistasis between FAM134B and CD39.
Genetic analysis of an allergic rhinitis cohort reveals an intercellular epistasis between FAM134B and CD39.
BMC medical genetics (2014-06-28)
Rossella Melchiotti, Kia Joo Puan, Anand Kumar Andiappan, Tuang Yeow Poh, Mireille Starke, Li Zhuang, Kerstin Petsch, Tuck Siong Lai, Fook Tim Chew, Anis Larbi, De Yun Wang, Michael Poidinger, Olaf Rotzschke
ABSTRACT
Extracellular ATP is a pro-inflammatory molecule released by damaged cells. Regulatory T cells (Treg) can suppress inflammation by hydrolysing this molecule via ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1), also termed as CD39. Multiple studies have reported differences in CD39+ Treg percentages in diseases such as multiple sclerosis, Hepatitis B and HIV-1. In addition, CD39 polymorphisms have been implicated in immune-phenotypes such as susceptibility to inflammatory bowel disease and AIDS progression. However none of the studies published so far has linked disease-associated variants with differences in CD39 Treg surface expression. This study aims at identifying variants affecting CD39 expression on Treg and at evaluating their association with allergic rhinitis, a disease characterized by a strong Treg involvement. Cohorts consisting of individuals of different ethnicities were employed to identify any association of CD39 variants to surface expression. Significant variant(s) were tested for disease association in a published GWAS cohort by one-locus and two-locus genetic analyses based on logistic models. Further functional characterization was performed using existing microarray data and quantitative RT-PCR on sorted cells. Our study shows that rs7071836, a promoter SNP in the CD39 gene region, affects the cell surface expression on Treg cells but not on other CD39+ leukocyte subsets. Epistasis analysis revealed that, in conjunction with a SNP upstream of the FAM134B gene (rs257174), it increased the risk of allergic rhinitis (P = 1.98 × 10-6). As a promoter SNP, rs257174 controlled the expression of the gene in monocytes but, notably, not in Treg cells. Whole blood transcriptome data of three large cohorts indicated an inverse relation in the expression of the two proteins. While this observation was in line with the epistasis data, it also implied that a functional link must exist. Exposure of monocytes to extracellular ATP resulted in an up-regulation of FAM134B gene expression, suggesting that extracellular ATP released from damaged cells represents the connection for the biological interaction of CD39 on Treg cells with FAM134B on monocytes. The interplay between promoter SNPs of CD39 and FAM134B results in an intercellular epistasis which influences the risk of a complex inflammatory disease.
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, HPLC Plus
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00