MilliporeSigma
  • Metabolomic discovery of novel urinary galabiosylceramide analogs as Fabry disease biomarkers.
Metabolomic discovery of novel urinary galabiosylceramide analogs as Fabry disease biomarkers.
Journal of the American Society for Mass Spectrometry (2015-01-15)
Michel Boutin, Christiane Auray-Blais
ABSTRACT
Fabry disease is an X-linked, complex, multisystemic lysosomal storage disorder presenting marked phenotypic and genotypic variability among affected male and female patients. Glycosphingolipids, mainly globotriaosylceramide (Gb(3)) isoforms/analogs, globotriaosylsphingosine (lyso-Gb(3)) and analogs, as well as galabiosylceramide (Ga(2)) isoforms/analogs accumulate in the vascular endothelium, nerves, cardiomyocytes, renal glomerular and tubular epithelial cells, and biological fluids. The search for biomarkers reflecting disease severity and progression is still on-going. A metabolomic study using quadrupole time-of-flight mass spectrometry has revealed 22 galabiosylceramide isoforms/analogs in urine of untreated Fabry patients classified in seven groups according to their chemical structure: (1) Saturated fatty acid; (2) one extra double bond; (3) two extra double bonds; (4) hydroxylated saturated fatty acid; (5) hydroxylated fatty acid and one extra double bond; (6) hydrated sphingosine and hydroxylated fatty acid; (7) methylated amide linkage. Relative quantification of both Ga(2) and Gb(3) isoforms/analogs was performed. All these biomarkers are significantly more abundant in urine samples from untreated Fabry males compared with healthy male controls. A significant amount of Ga(2) isoforms/analogs, accounting for 18% of all glycosphingolipids analyzed (Ga(2) + Gb(3) and respective isoforms/analogs), were present in urine of Fabry patients. Gb(3) isoforms containing saturated fatty acids are the most abundant (60.9%) compared with 26.3% for Ga(2). A comparison between Ga(2) isoforms/analogs and their Gb(3) counterparts also showed that the proportion of analogs with hydroxylated fatty acids is significantly greater for Ga(2) (35.8%) compared with Gb(3) (1.9%). These results suggest different biological pathways involved in the synthesis and/or degradation of Gb(3) and Ga(2) metabolites.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Terfenadine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Terfenadine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in ethanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M in ethanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.02 M in ethanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in isopropanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00