MilliporeSigma
  • Disadvantaged pre-schoolers attending day care in Salvador, Northeast Brazil have a low prevalence of anaemia and micronutrient deficiencies.
Disadvantaged pre-schoolers attending day care in Salvador, Northeast Brazil have a low prevalence of anaemia and micronutrient deficiencies.
Public health nutrition (2013-09-07)
Rebecca L Lander, Karl B Bailey, Alastair G Lander, Abdulmonem A Alsaleh, Hugo C Costa-Ribeiro, Angela P Mattos, Danile L Barreto, Lisa A Houghton, Ian M Morison, Sheila M Williams, Rosalind S Gibson
ABSTRACT
To examine the micronutrient status of disadvantaged pre-schoolers from Northeast Brazil, following the introduction of pro-poor policies, by assessing the prevalence of anaemia and micronutrient deficiencies and the role of sociodemographic factors, genetic Hb disorders and parasitic infections. In a cross-sectional study, data on sociodemographic status, health, growth, genetic Hb disorders, parasites and nutrient supply from day-care meals were obtained. Fasting blood samples were collected and analysed for Hb, serum ferritin, transferrin receptor, folate, vitamin B₁₂, retinol, Zn and Se. Seven philanthropic day-care centres serving urban slums in Salvador, Northeast Brazil. Pre-schoolers aged 3-6 years from disadvantaged households. Of the 376 sampled children, 94 % were of black or mixed race; 33 % and 29 % had at least one genetic Hb disorder and intestinal parasite, respectively. Stunting and underweight were ≤5 %; 14 % were overweight. Day-care centres supplied micronutrient-dense meals and snacks each weekday. Less than 10 % of pre-schoolers had anaemia and micronutrient deficiencies. Predictors (P < 0·05) of Hb were α(3·7) thalassaemia, Se and retinol (but not ferritin). Micronutrient predictors (P < 0·05) were: elevated α₁-glycoprotein for ferritin, Hb AS and BMI Z-score >1 for transferrin receptor, Zn and elevated α₁-glycoprotein for retinol, sex and helminths for Se, helminths for vitamin B₁₂, and Giardia intestinalis infection for serum folate. Impaired growth, anaemia and micronutrient deficiencies were uncommon among these disadvantaged pre-schoolers attending day care. A range of interventions including provision of micronutrient-dense, fortified day-care meals, deworming and vitamin A supplementation likely contributed to improved micronutrient status, suggesting expanded coverage of these programmes.
Product Description
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%