MilliporeSigma
  • Preparation and chromatographic performance evaluation of cucurbit[7]uril immobilized silica.
Preparation and chromatographic performance evaluation of cucurbit[7]uril immobilized silica.
Journal of chromatography. A (2014-12-22)
Liyun Ma, Si-Min Liu, Lin Yao, Li Xu
ABSTRACT
Cucurbit[7]uril (CB7), one host macromolecule, possesses a hydrophobic inner cavity and two identical hydrophilic carbonyl fringe portals, thus interacting with a variety of guest molecules through hydrophobic, hydrogen bonding and ion-dipole interactions. These characteristics make CB7 a good candidate as the stationary phase (SP) modifier in high performance liquid chromatography (HPLC). In the present work, CB7 was immobilized onto silica (CB7-SiO2) by a sol-gel approach. The obtained CB7-SiO2 was characterized by Fourier transform infrared spectroscopy, thermogravimetric analysis and elemental analysis. The chromatographic performance of CB7-SiO2 was then studied in detail, in both reversed phase (RP) and hydrophilic interaction liquid chromatographic (HILIC) modes. For probe aromatic compounds and dyes, the typical RP behavior was observed, mainly due to the predominant hydrophobic interaction between the analytes and CB7-SiO2. For probe alkaloids, the "U-shape" curves were obtained; in the methanol-rich part, HILIC performance was obvious, probably owing to hydrogen bonding and ion-dipole interactions. Compared to bare SiO2 SP, the stronger retention and different selectivity were observed on CB7-SiO2. The CB7-SiO2 SP remained stable within at least 11,000 column volumes of the MP, revealing the satisfactory stability of it. The proposed method integrated the advantages of CB7 and sol-gel method provided a feasible way for the wider application of CBs in separation science.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Potassium bromide, JIS special grade, 99.0-100.2%
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ethanol, 94.8-95.8%
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Supelco
Methanol, analytical standard
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)