MilliporeSigma
  • Kidney-specific drug delivery system for renal fibrosis based on coordination-driven assembly of catechol-derived chitosan.
Kidney-specific drug delivery system for renal fibrosis based on coordination-driven assembly of catechol-derived chitosan.
Biomaterials (2014-05-28)
Hongzhi Qiao, Minjie Sun, Zhigui Su, Ying Xie, Minglei Chen, Li Zong, Yahan Gao, Huipeng Li, Jianping Qi, Qun Zhao, Xiaochen Gu, Qineng Ping
ABSTRACT
Renal fibrosis is a common progressive kidney disease, and there is a lack of efficient treatment for the condition. In this study, we designed a kidney-specific nanocomplex by forming coordination-driven assembly from catechol-derived low molecular weight chitosan (HCA-Chi), metal ions and active drug molecules. The coordination activities of various metals and ligands, cytotoxicity, immunogenicity and biodistribution of HCA-Chi were investigated. Autofluorescent doxorubicin (DOX) was selected to fabricate HCA-Chi-Cu-DOX ternary nanocomplex for investigating cellular uptake behavior, transmembrane and targeting properties. The nanodevice demonstrated satisfactory stability under normal physiological conditions and pH-responsive drug release in acidic environments. Uptake of HCA-Chi-Cu-DOX by HK-2 cells was dependent on exposure time, concentration, and temperature, and was inhibited by blockers of megalin receptor. Tissue distribution showed that HCA-Chi-Cu-DOX nanocomplex was specifically accumulated in kidney with a renal relative uptake rate (r(e)) of 25.6. When active anti-fibrosis compound emodin was installed in HCA-Chi-Zn-emodin and intravenously injected to the ureter obstructed mice, obvious attenuation of fibrotic progression was exhibited. It was concluded that HCA-Chi coordination-driven nanocomplex showed special renal targeting capacity and could be utilized to develop drug delivery systems for treating renal fibrosis.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Millipore
TDA Reagent, suitable for microbiology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00