MilliporeSigma
  • Evaluation of efficacy of natural astaxanthin and vitamin E in prevention of colistin-induced nephrotoxicity in the rat model.
Evaluation of efficacy of natural astaxanthin and vitamin E in prevention of colistin-induced nephrotoxicity in the rat model.
Environmental toxicology and pharmacology (2014-04-09)
Zohra Ghlissi, Ahmed Hakim, Assaad Sila, Hela Mnif, Khaled Zeghal, Tarek Rebai, Ali Bougatef, Zouheir Sahnoun
ABSTRACT
We evaluated the effect of astaxanthin (ASX) and vitamin E (vit E) on colistin methanesulfonate (CMS) induced-nephrotoxicity in rats. Animals were treated with sterile saline, 300000 or 450 000 IU/kg/day of CMS, CMS + ASX (20 mg/kg), CMS + vit E (100 mg/kg), or CMS + 1 ml/kg olive oil (OO) for 7 days. The plasma/urine creatinine (Cr) level, urine γ-glutamyl-transferase (GGT) level, and renal tissue activities in malondialdehyde (MDA), superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx) and reductase (GSH), as well as renal histology were performed. CMS induced a tubular damage, increased the GGT and MDA levels, and decreased the activities of SOD, CAT, GPx and GSH. Co-treatment with ASX or vit E restored all biochemical parameters cited above and improved the histopathological damage. Nephrotoxicity induced by CMS might be due to oxidative damage. The improvement by ASX or vit E seems to be related to their antioxidant properties.
Sigma-Aldrich
α-Tocopherol, ≥95.5%
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard