MilliporeSigma
  • Potential of syncytiotrophoblasts isolated from the cervical mucus for early non-invasive prenatal diagnosis: evidence of a vanishing twin.
Potential of syncytiotrophoblasts isolated from the cervical mucus for early non-invasive prenatal diagnosis: evidence of a vanishing twin.
Clinica chimica acta; international journal of clinical chemistry (2014-09-15)
Debbie Mantzaris, David S Cram
ABSTRACT
Non-invasive methods to assess the foetal genome during pregnancy will provide new opportunities to offer pregnant women a more comprehensive genetic diagnosis of their established foetus. The aim of this study was to determine the presence and frequency of foetal cells in transcervical cell (TCC) mucus samples from pregnant women and determine their suitability for early prenatal diagnosis. Syncytiotrophoblasts in aspirated TCC mucus samples were identified by immunostaining with the foetal-specific antibody NDOG1. Genetic analysis of foetal cells was performed by laser capture microdissection and quantitative fluorescent PCR (QF-PCR). In 116 of 207 (56%) TCC samples, abundant syncytiotrophoblasts were retrieved. However, when TCC samples were stratified for the presence of chorionic villous fragments, syncytiotrophoblasts were identified in 85 of 109 (78%) samples. Significant numbers of syncytiotrophoblasts were found in TCC samples collected between 6 and 9weeks of gestation (mean 741, range 25-2884). QF-PCR analysis of NDOG1 positive syncytiotrophoblasts and matching maternal DNA confirmed their foetal origin and correct foetal cell sexing was achieved in 97% of TCC samples. The one discordant sex diagnosis was associated with a dizygotic dichorionic twin pregnancy resulting from the implantation of a female T21 embryo and a normal male embryo, where the female T21 foetus had succumbed at 6weeks of gestation and was vanishing. Syncytiotrophoblasts can be successfully isolated from TCC samples and represent a suitable source of cells for genetic analysis of the established foetus in early pregnancy. The study highlights a vanishing twin as a potential cause for discordant non-invasive prenatal test results.
Product Description
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)