MilliporeSigma
  • Design of lipotomes as a novel dual functioning nanocarrier for bioavailability enhancement of lacidipine: in-vitro and in-vivo characterization.
Design of lipotomes as a novel dual functioning nanocarrier for bioavailability enhancement of lacidipine: in-vitro and in-vivo characterization.
International journal of pharmaceutics (2014-07-01)
Nermeen Adel ElKasabgy, Ibrahim Elsayed, Ahmed Hassen Elshafeey
ABSTRACT
Lipotomes were designed to enhance lacidipine's oral bioavailability by improving its solubility and enhancing the oral lymphatic uptake. Lipotomes were prepared using cetyl alcohol and Tween(®) 80 using a thin film hydration technique. Cetyl alcohol was chosen for imparting a lipophilic environment that would enforce the lymphatic uptake while Tween(®) 80 would improve drug solubility within the lipotomes. Lipotomes were characterized by analyzing their particle size, solubilization efficiency and in-vitro drug release. Central composite design was applied to statistically optimize the formulations using Design-Expert(®) software. The optimum formula (OLT) was made up of excipients:drug ratio of 36.59:1 w/w and Tween(®) 80:cetyl alcohol ratio of 4:1 w/w. OLT was lyophilized and filled into Eudragit(®) L100 enteric coated capsules. Mannitol (10% w/v) was the ideal cryoprotectant to retain the physicochemical characteristics of the OLT formulation after lyophilization. In conclusion, the selected lyophilized formula (L3) succeeded in enhancing drug's oral bioavailability in human volunteers compared to the commercial product confirming the success of lipotomes as a novel oral nanocarrier for insoluble drugs having extensive first pass metabolism.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Trehalose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
tert-Butyl methyl ether, analytical standard
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material