MilliporeSigma
Absolute quantification method for protein concentration.
Analytical chemistry (2014-11-21)
Mingdong Li, Jiaojie Tan, Michael J Tarlov, Michael R Zachariah
ABSTRACT
A fast and accurate assay to determine the absolute concentration of proteins is described based on direct measurement of droplet entrapped oligomer formation in electrospray. Here we demonstrate the approach using electrospray differential mobility analysis (ES-DMA), which can distinguish monomers and dimers from higher order oligomers. A key feature of the method is that it allows determination of the absolute number concentration of proteins eliminating the need for protein-specific calibration. The method was demonstrated by measuring the concentration of a NIST Standard Reference Material 927e (bovine serum albumin), a high-purity immunoglobulin G 1κ, and a formulated Rituximab. The method may be applied to any electrospray source, regardless of diagnostic tool (e.g., MS or ion-mobility, etc.), provided the electrospray is operated in a droplet-fission mode.
Sigma-Aldrich
Sucrose, JIS special grade
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, SAJ first grade
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)