MilliporeSigma
  • Plasmid-encoded tetracycline efflux pump protein alters bacterial stress responses and ecological fitness of Acinetobacter oleivorans.
Plasmid-encoded tetracycline efflux pump protein alters bacterial stress responses and ecological fitness of Acinetobacter oleivorans.
PloS one (2014-09-18)
Hyerim Hong, Jaejoon Jung, Woojun Park
ABSTRACT
Acquisition of the extracellular tetracycline (TC) resistance plasmid pAST2 affected host gene expression and phenotype in the oil-degrading soil bacterium, Acinetobacter oleivorans DR1. Whole-transcriptome profiling of DR1 cells harboring pAST2 revealed that all the plasmid genes were highly expressed under TC conditions, and the expression levels of many host chromosomal genes were modulated by the presence of pAST2. The host energy burden imposed by replication of pAST2 led to (i) lowered ATP concentrations, (ii) downregulated expression of many genes involved in cellular growth, and (iii) reduced growth rate. Interestingly, some phenotypes were restored by deleting the plasmid-encoded efflux pump gene tetH, suggesting that the membrane integrity changes resulting from the incorporation of efflux pump proteins also resulted in altered host response under the tested conditions. Alteration of membrane integrity by tetH deletion was shown by measuring permeability of fluorescent probe and membrane hydrophobicity. The presence of the plasmid conferred peroxide and superoxide resistance to cells, but only peroxide resistance was diminished by tetH gene deletion, suggesting that the plasmid-encoded membrane-bound efflux pump protein provided peroxide resistance. The downregulation of fimbriae-related genes presumably led to reduced swimming motility, but this phenotype was recovered by tetH gene deletion. Our data suggest that not only the plasmid replication burden, but also its encoded efflux pump protein altered host chromosomal gene expression and phenotype, which also alters the ecological fitness of the host in the environment.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Pricing and availability is not currently available.
Supelco
Hexadecane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Pricing and availability is not currently available.