MilliporeSigma
  • Phosphodiesterase 5 inhibition ameliorates angiontensin II-induced podocyte dysmotility via the protein kinase G-mediated downregulation of TRPC6 activity.
Phosphodiesterase 5 inhibition ameliorates angiontensin II-induced podocyte dysmotility via the protein kinase G-mediated downregulation of TRPC6 activity.
American journal of physiology. Renal physiology (2014-04-18)
Gentzon Hall, Janelle Rowell, Federica Farinelli, Rasheed A Gbadegesin, Peter Lavin, Guanghong Wu, Alison Homstad, Andrew Malone, Thomas Lindsey, Ruiji Jiang, Robert Spurney, Gordon F Tomaselli, David A Kass, Michelle P Winn
ABSTRACT
The emerging role of the transient receptor potential cation channel isotype 6 (TRPC6) as a central contributor to various pathological processes affecting podocytes has generated interest in the development of therapeutics to modulate its function. Recent insights into the regulation of TRPC6 have revealed PKG as a potent negative modulator of TRPC6 conductance and associated signaling via its phosphorylation at two highly conserved amino acid residues: Thr(69)/Thr(70) (Thr(69) in mice and Thr(70) in humans) and Ser(321)/Ser(322) (Ser(321) in mice and Ser(322) in humans). Here, we tested the role of PKG in modulating TRPC6-dependent responses in primary and conditionally immortalized mouse podocytes. TRPC6 was phosphorylated at Thr(69) in nonstimulated podocytes, but this declined upon ANG II stimulation or overexpression of constitutively active calcineurin phosphatase. ANG II induced podocyte motility in an in vitro wound assay, and this was reduced 30-60% in cells overexpressing a phosphomimetic mutant TRPC6 (TRPC6T70E/S322E) or activated PKG (P < 0.05). Pretreatment of podocytes with the PKG agonists S-nitroso-N-acetyl-dl-penicillamine (nitric oxide donor), 8-bromo-cGMP, Bay 41-2772 (soluble guanylate cyclase activator), or phosphodiesterase 5 (PDE5) inhibitor 4-{[3',4'-(methylenedioxy)benzyl]amino}[7]-6-methoxyquinazoline attenuated ANG II-induced Thr(69) dephosphorylation and also inhibited TRPC6-dependent podocyte motility by 30-60%. These data reveal that PKG activation strategies, including PDE5 inhibition, ameliorate ANG II-induced podocyte dysmotility by targeting TRPC6 in podocytes, highlighting the potential therapeutic utility of these approaches to treat hyperactive TRPC6-dependent glomerular disease.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt, ≥95% (HPLC), powder