MilliporeSigma
  • Enhanced differentiation of neural progenitor cells into neurons of the mesencephalic dopaminergic subtype on topographical patterns.
Enhanced differentiation of neural progenitor cells into neurons of the mesencephalic dopaminergic subtype on topographical patterns.
Biomaterials (2015-01-17)
Kenneth K B Tan, Jason Y Tann, Sharvari R Sathe, Seok Hong Goh, Dongliang Ma, Eyleen L K Goh, Evelyn K F Yim
ABSTRACT
Parkinson's disease (PD) is a neurodegenerative disease attributed to the loss of midbrain dopaminergic (DA) neurons. The current lack of predictive models for this disease has been hampered by the acquirement of robust cells, posing a major barrier to drug development. Differentiation of stem cells into subtype specific cells may be guided by appropriate topographical cues but the role of topography has hitherto not been well understood. We used a Multi-Architecture (MARC) chip with various topographical structures and identified three topographies, which generate DA neurons from murine hippocampal neural progenitor cells with the highest percentage of neuronal (β-III-tubulin positive) and dopaminergic (tyrosine hydroxylase positive) populations. Analysis on single pattern structures showed that 2 μm gratings with 2 μm spacing and 2 μm height (2 μm gratings) and 2 μm gratings with hierarchical structure produced cells with the highest gene expression of TH and PITX3, with the longest neurite and highest percentage of alignment. Quantitative image analysis showed the 2 μm gratings produced cells with the highest expression of pituitary homeobox 3 (PITX3), LIM homeobox transcription factor 1 alpha (LMX1a), aldehyde dehydrogenase 1 family member A1 (ALDH1a1) and microtubule associated protein 2 (MAP2), as compared to nano-gratings and unpatterned controls. These patterns also enhance DA neuron differentiation on different substrate rigidities, as seen on both poly-dimethylsiloxane (PDMS) and tissue culture polystyrene (TCPS) substrates. These results show the use of topographical influence for neuronal subtype specification, which could be translated into a wide range of clinical applications for PD.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Streptomycin sulfate salt, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Pricing and availability is not currently available.
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Pricing and availability is not currently available.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and availability is not currently available.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.