MilliporeSigma
  • A novel technology for multiplex gene expression analysis directly from whole blood samples stabilized at ambient temperature using an RNA-stabilizing buffer.
A novel technology for multiplex gene expression analysis directly from whole blood samples stabilized at ambient temperature using an RNA-stabilizing buffer.
The Journal of molecular diagnostics : JMD (2015-02-17)
Chang Hee Kim, Majid Abedi, Yenbou Liu, Sree Panuganti, Francisco Flores, Kevin R Shah, Hannah Catterall, Krishna S Morampudi, Robert Terbrueggen
ABSTRACT
We describe a novel method, based on target-dependent chemical ligation of probes, which simplifies the multiplexed quantitation of gene expression from blood samples by eliminating the RNA purification step. Gene expression from seven genes was evaluated over a range of sample inputs (16.7 to 0.25 μL of whole blood in serial dilutions) from three healthy donors. Mean CVs were ≤11% for five technical replicates for whole blood inputs ≥2.1 μL. The method showed a limit of detection of 300 copies of RNA by using titration of in vitro transcripts for four genes. Gene expression measured on stabilized blood samples was highly correlated (Spearman rank correlation method, ρ = 0.80) to gene expression results obtained with RNA isolated from matched samples (three donors, five technical replicates). Gene expression changes determined with seven radiation-responsive genes on six healthy donor blood samples before and after ex vivo irradiation were highly correlated (ρ = 0.93) to those measured with a TaqMan quantitative real-time RT-PCR assay on RNA purified from matched samples. Thus, this method is reproducible, sensitive, and correlated to quantitative real-time RT-PCR and may be used to streamline the multiplex gene expression analysis of large numbers of stabilized blood samples without RNA purification.
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.