MilliporeSigma
  • Coimmobilization of l-methioninase and glutamate dehydrogenase: Novel approach for L-homoalanine synthesis.
Coimmobilization of l-methioninase and glutamate dehydrogenase: Novel approach for L-homoalanine synthesis.
Biotechnology and applied biochemistry (2014-10-03)
Ashraf S A El-Sayed, Marwa A Yassin, Hend Ibrahim
ABSTRACT
L-Homoalanine, a nonnatural amino acid that is rarely found in human and microorganisms, is used in the synthesis of various medically pivotal antiepileptic drugs and antituberculosis compounds. l-Homoalanine can be synthesized by different enzymatic approaches. In this article, the synthesis of l-homoalanine from l-methionine was explored by coimmobilization of Aspergillus flavipes l-methioninase (AfMETase) and glutamate dehydrogenase (GDH) on polyacrylamide and chitosan. Polyacrylamide coimmobilized AfMETase and GDH displayed a maximum reactivity for the synthesis of homoalanine from l-methionine. The chitosan-coimmobilized AfMETase and GDH retain about 70% of their initial activity of l-homoalanine production by the fifth catalytic reusability cycle as compared with 50% for polyacrylamide coimmobilizate. Catalytic conditions were optimized for the maximum yield of homoalanine. Homoalanine was purified by cationic and anionic chromatographs and the proton nuclear magnetic resonance (H-NMR) analysis of the lyophilized sample displayed a unique chemical structure identical to the authentic homoalanine. Using dependable dual action of AfMETase and GDH immobilized on a solid support is a novel approach for in vitro enzymatic synthesis of l-homoalanine from l-methionine, and the immobilized enzymes can be reused many times without any significant loss of their activities.
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
SAFC
L-Methionine
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
L-Methionine, SAJ special grade, ≥98.5%